missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CHST12 Partial ORF (NP_061111.1, 56 a.a. - 150 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055501-Q02.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16025565

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

Chondroitin 4-sulfotransferases, such as CHST12, add sulfate to position 4 of N-acetylgalactosamine residues in chondroitin (Hiraoka et al., 2000 [PubMed 10781601]).[supplied by OMIM]

Sequence: RDRELTADSDVDEFLDKFLSAGVKQSDLPRKETEQPPAPGSMEESVRGYDWSPRDARRSPDQGRQQAERRSVLRGFCANSSLAFPTKERAFDDIP
Specifications

Specifications

NP_061111.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.19kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RDRELTADSDVDEFLDKFLSAGVKQSDLPRKETEQPPAPGSMEESVRGYDWSPRDARRSPDQGRQQAERRSVLRGFCANSSLAFPTKERAFDDIP
RUO
CHST12
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
55501
CHST12 (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C4S-2/C4ST-2/C4ST2
CHST12
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers