Learn More
R&D Systems™ Human Chromogranin A DuoSet ELISA
Click to view available options
Sufficient For:
Five 96-well plates
Unit Size:
1 set
Description
The Human Chromogranin A DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Chromogranin A.
Specifications
Specifications
| Assay Range | 62.50 to 4,000 pg/mL |
| Immunoassay Kit Format | 96-well Plate |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
| Sufficient For | Five 96-well plates |
| Target Species | Human |
| Gene ID (Entrez) | 1113 |
| Kit Contents | 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Target | Chromogranin A |