missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CHMP5 Full-length ORF (NP_057494.2, 1 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051510-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16190083

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

CHMP5 belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A (MIM 164010), is required for both MVB formation and regulation of cell cycle progression (Tsang et al., 2006 [PubMed 16730941]).[supplied by OMIM]

Sequence: MNRLFGKAKPKAPPPSLTGCIGTVDSRAESIDKKISRLDAELVKYKDQIKKMREGPAKNMVKQKALRVLKQKRMYEQQRDNLAQQSFNMEQANYTIQSLKDTKTTVDAMKLGVKEMKKAYKQVKIDQIEDLQDQLEDMMEDANEIQEALSRSYGTPELDEDDLEAELDALGDELLADEDSSYLDEAASAPAIPEGVPTDTKNKDGVLVDEFGLPQIPAS
Specifications

Specifications

NP_057494.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C9orf83/CGI-34/HSPC177/PNAS-2/SNF7DC2
CHMP5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
51510
CHMP5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNRLFGKAKPKAPPPSLTGCIGTVDSRAESIDKKISRLDAELVKYKDQIKKMREGPAKNMVKQKALRVLKQKRMYEQQRDNLAQQSFNMEQANYTIQSLKDTKTTVDAMKLGVKEMKKAYKQVKIDQIEDLQDQLEDMMEDANEIQEALSRSYGTPELDEDDLEAELDALGDELLADEDSSYLDEAASAPAIPEGVPTDTKNKDGVLVDEFGLPQIPAS
RUO
CHMP5
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers