missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CHIC2 Partial ORF (NP_036242, 21 a.a. - 84 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026511-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105526

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the CHIC family of proteins. The encoded protein contains a cysteine-rich hydrophobic (CHIC) motif, and is localized to vesicular structures and the plasma membrane. This gene is associated with some cases of acute myeloid leukemia. [provided by RefSeq]

Sequence: QLLKYSPDPVVVRGSGHVTVFGLSNKFESEFPSSLTGKVAPEEFKASINRVNSCLKKNLPVNVR
Specifications

Specifications

NP_036242
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.78kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QLLKYSPDPVVVRGSGHVTVFGLSNKFESEFPSSLTGKVAPEEFKASINRVNSCLKKNLPVNVR
RUO
CHIC2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
26511
CHIC2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BTL/MGC21173
CHIC2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers