missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CHD1 Partial ORF (NP_001261, 1177 a.a. - 1272 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001105-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16156264

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

The CHD family of proteins is characterized by the presence of chromo (chromatin organization modifier) domains and SNF2-related helicase/ATPase domains. CHD genes alter gene expression possibly by modification of chromatin structure thus altering access of the transcriptional apparatus to its chromosomal DNA template. [provided by RefSeq]

Sequence: IKALKDSSSGTERTGGRLGKVKGPTFRISGVQVNAKLVISHEEELIPLHKSIPSDPEERKQYTIPCHTKAAHFDIDWGKEDDSNLLIGIYEYGYGS
Specifications

Specifications

NP_001261
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.3kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IKALKDSSSGTERTGGRLGKVKGPTFRISGVQVNAKLVISHEEELIPLHKSIPSDPEERKQYTIPCHTKAAHFDIDWGKEDDSNLLIGIYEYGYGS
RUO
CHD1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1105
CHD1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686E2337
CHD1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers