Learn More
Abnova™ Human Chac1 Full-Length Orf (Np_077016.1, 1 A.A.-222 A.A.) Recombinant Protein With Gst Tag At N-Terminal.
Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.
Brand: Abnova™ H00079094-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Good Folding
- Excellent amino acids labeling
Immunogen Sequence: MKQESAAPNTPPTSQSPTPSAQFPRNDGDPQALWIFGYGSLVWRPDFAYSDSRVGFVRGYSRRFWQGDTFHRGSDKMPGRVVTLLEDHEGCTWGVAYQVQGEQVSKALKYLNVREAVLGGYDTKEVTFYPQDAPDQPLKALAYVAT
Specifications
NP_077016.1 | |
Liquid | |
79094 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC4504 | |
CHAC1 |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. | |
CHAC1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MKQESAAPNTPPTSQSPTPSAQFPRNDGDPQALWIFGYGSLVWRPDFAYSDSRVGFVRGYSRRFWQGDTFHRGSDKMPGRVVTLLEDHEGCTWGVAYQVQGEQVSKALKYLNVREAVLGGYDTKEVTFYPQDAPDQPLKALAYVATPQNPGYLGPAPEEAIATQILACRGFSGHNLEYLLRLADFMQLCGPQAQDEHLAAIVDAVGTMLPCFCPTEQALALV | |
RUO | |
CHAC1 | |
GST |
For Research Use Only