missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CFD Partial ORF (NP_001919.2, 154 a.a. - 253 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_001919.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1675 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16167154
|
Abnova™
H00001675-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16157154
|
Abnova™
H00001675-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a member of the trypsin family of peptidases. The encoded protein is a component of the alternative complement pathway best known for its role in humoral suppression of infectious agents. This protein is also a serine protease that is secreted by adipocytes into the bloodstream. Finally, the encoded protein has a high level of expression in fat, suggesting a role for adipose tissue in immune system biology. [provided by RefSeq]
Sequence: GIVNHAGRRPDSLQHVLLPVLDRATCNRRTHHDGAITERLMCAESNRRDSCKGDSGGPLVCGGVLEGVVTSGSRVCGNRKKPGIYTRVASYAAWIDSVLASpecifications
NP_001919.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ADIPSIN/ADN/DF/PFD | |
CFD | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1675 | |
CFD (Human) Recombinant Protein (Q01) | |
GIVNHAGRRPDSLQHVLLPVLDRATCNRRTHHDGAITERLMCAESNRRDSCKGDSGGPLVCGGVLEGVVTSGSRVCGNRKKPGIYTRVASYAAWIDSVLA | |
RUO | |
CFD | |
Wheat Germ (in vitro) | |
GST | |
Liquid |