missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CEP57 Full-length ORF (NP_055494.2, 1 a.a. - 500 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009702-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16104022

  • 335.00€ / 10µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

Translokin binds basic fibroblast growth factor (FGF2; MIM 134920) and mediates its nuclear translocation and mitogenic activity (Bossard et al., 2003 [PubMed 12717444]).[supplied by OMIM]

Sequence: MAAASVSAASGSHLSNSFAEPSRSNGSMVRHSSSPYVVYPSDKPFLNSDLRRSPSKPTLAYPESNSRAIFSALKNLQDKIRRLELERIQAEESVKTLSRETIEYKKVLDEQIQERENSKNEESKHNQELTSQLLAAENKCNLLEKQLEYMRNMIKHAEMERTSVLEKQVSLERERQHDQTHVQSQLEKLDLLEQEYNKLTTMQALAEKKMQELEAKLHEEEQERKRMQAKAAELQTGLETNRLIFEDKATPCVPNARRIKKKKSKPPEKKSSRNYFGAQPHYRLCLGDMPFVAGKSTSPSHAVVANVQLVLHLMKQHSKALCNDRVINSIPLAKQVSSRGGKSKKLSVTPPSSNGINEELSEVLQTLQDEFGQMSFDHQQLAKLIQESPTVELKDKLECELEALVGRMEAKANQITKVRKYQAQLEKQKLEKQKKELKATKKTLDEERNSSSRSGITGTTNKKDFMKLRPGEKRRKNLQLLKDMQSIQNSLQSSSLCWDY
Specifications

Specifications

NP_055494.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
80.63kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0092/PIG8/TSP57
CEP57
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9702
CEP57 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAASVSAASGSHLSNSFAEPSRSNGSMVRHSSSPYVVYPSDKPFLNSDLRRSPSKPTLAYPESNSRAIFSALKNLQDKIRRLELERIQAEESVKTLSRETIEYKKVLDEQIQERENSKNEESKHNQELTSQLLAAENKCNLLEKQLEYMRNMIKHAEMERTSVLEKQVSLERERQHDQTHVQSQLEKLDLLEQEYNKLTTMQALAEKKMQELEAKLHEEEQERKRMQAKAAELQTGLETNRLIFEDKATPCVPNARRIKKKKSKPPEKKSSRNYFGAQPHYRLCLGDMPFVAGKSTSPSHAVVANVQLVLHLMKQHSKALCNDRVINSIPLAKQVSSRGGKSKKLSVTPPSSNGINEELSEVLQTLQDEFGQMSFDHQQLAKLIQESPTVELKDKLECELEALVGRMEAKANQITKVRKYQAQLEKQKLEKQKKELKATKKTLDEERNSSSRSGITGTTNKKDFMKLRPGEKRRKNLQLLKDMQSIQNSLQSSSLCWDY
RUO
CEP57
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers