missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CENTB2 Partial ORF (NP_036419, 473 a.a. - 581 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023527-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: DVINRVYEANVEKMGIKKPQPGQRQEKEAYIRAKYVERKFVDKYSISLSPPEQQKKFVSKSSEEKRLSISKFGPGDQVRASAQSSVRSNDSGIQQSSDDGRESLPSTVSSpecifications
NP_036419 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DVINRVYEANVEKMGIKKPQPGQRQEKEAYIRAKYVERKFVDKYSISLSPPEQQKKFVSKSSEEKRLSISKFGPGDQVRASAQSSVRSNDSGIQQSSDDGRESLPSTVS | |
RUO | |
ACAP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23527 | |
CENTB2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CENTB2/CNT-B2/KIAA0041 | |
ACAP2 | |
Recombinant | |
wheat germ expression system |