missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CELSR2 Partial ORF (NP_001399, 124 a.a. - 233 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001952-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194401

  • 528.00€ / 25µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a member of the flamingo subfamily, part of the cadherin superfamily. The flamingo subfamily consists of nonclassic-type cadherins; a subpopulation that does not interact with catenins. The flamingo cadherins are located at the plasma membrane and have nine cadherin domains, seven epidermal growth factor-like repeats and two laminin A G-type repeats in their ectodomain. They also have seven transmembrane domains, a characteristic unique to this subfamily. It is postulated that these proteins are receptors involved in contact-mediated communication, with cadherin domains acting as homophilic binding regions and the EGF-like domains involved in cell adhesion and receptor-ligand interactions. The specific function of this particular member has not been determined. [provided by RefSeq]

Sequence: SPQGKLTLPEEHPCLKAPRLRCQSCKLAQAPGLRAGERSPEESLGGRRKRNVNTAPQFQPPSYQATVPENQPAGTPVASLRAIDPDEGEAGRLEYTMDALFDSRSNQFFS
Specifications

Specifications

NP_001399
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SPQGKLTLPEEHPCLKAPRLRCQSCKLAQAPGLRAGERSPEESLGGRRKRNVNTAPQFQPPSYQATVPENQPAGTPVASLRAIDPDEGEAGRLEYTMDALFDSRSNQFFS
RUO
CELSR2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1952
CELSR2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CDHF10/EGFL2/FLJ34118/FLJ42737/FLJ45143/FLJ45845/Flamingo1/KIAA0279/MEGF3
CELSR2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers