Learn More
Abcam Human CELF1 (CUG-BP1) knockout HEK-293T cell line
Description
CELF1 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 4 bp deletion in exon 2 and 5 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Quantity | 2 x 1,000,000 Cells |
| Regulatory Status | RUO |