Learn More
ACROBiosystems Human CEACAM-1 / CD66a Protein, His Tag
Brand: ACROBiosystems CE1-H5220-1MG
Additional Details : Weight : 0.00001kg
Description
Carcinoembryonic antigen-related cell adhesion molecule 1 (CEACAM1) is also known as Biliary glycoprotein 1 (BGP1), CD66a, which belongs to the immunoglobulin superfamily or CEA family. CEACAM1 /CD66a contains three Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. CEACAM1 /CD66a was described as an adhesion molecule mediating cell adhesion via both homophilic and heterophilic manners, and was detected on leukocytes, epithelia, and endothelia. Studies have revealed that CEACAM1 / BGP-1 performs actions in multiple cellular processes including tissue differentiation, angiogenesis, apoptosis, metastasis, as well as the modulation of innate and adaptive immune responses.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 44.1 kDa. The protein migrates as 57-100 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.Specifications
AAH14473 | |
1 mg | |
Gln 35 - Gly 428 | |
CEACAM1,CD66a,BGP,BGP1,BGPI | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
44.1 kDa | |
Human CEACAM-1, His Tag (CE1-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gly 428 (Accession # AAH14473). | |
-20°C | |
CEACAM-1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |