Learn More
Abnova™ Human CDH6 Full-length ORF (AAH00019.1, 1 a.a. - 663 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001004-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a type II classical cadherin from the cadherin superfamily. The encoded membrane protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Cadherins mediate cell-cell binding in a homophilic manner, contributing to the sorting of heterogeneous cell types and the maintenance of orderly structures such as epithelium. Strong transcriptional expression of this gene has been observed in hepatocellular and renal carcinoma cell lines, suggesting a possible role in metastasis and invasion. [provided by RefSeq]
Sequence: MRTYRYFLLLFWVGQPYPTLSTPLSKRTSGFPAKKRALELSGNSKNELNRSKRSWMWNQFFLLEEYTGSDYQYVGKLHSDQDRGDGSLKYILSGDGAGDLFIINENTGDIQATKRLDREEKPVYILRAQAINRRTGRPVEPESEFIIKIHDINDNEPIFTKEVYTATVPEMSDVGTFVVQVTATDADDPTYGNSAKVVYSILQGQPYFSVESETGIIKTALLNMDRENREQYQVVIQAKDMGGQMGGLSGTTTVNITLTDVNDNPPRFPQSTYQFKTPESSPPGTPIGRIKASDADVGENAEIEYSITDGEGLDMFDVITDQETQEGIITVKKLLDFEKKKVYTLKVEASNPYVEPRFLYLGPFKDSATVRIVVEDVDEPPVFSKLAYILQIREDAQINTTIGSVTAQDPDAARNPVKYSVDRHTDMDRIFNIDSGNGSIFTSKLLDRETLLWHNITVIATEINNPKQSSRVPLYIKVLDVNDNAPEFAEFYETFVCEKAKADQLIQTLHAVDKDDPYSGHQFSFSLAPEAASGSNFTIQDNKDNTAGILTRKNGYNRHEMSTYLLPVVISDNDYPVQSSTGTVTVRVCACDHHGNMQSCHAEALIHPTGLSTGALVAILLCIVILLGKLVLPASYLPMVRGSHCYCDTLDLSASPIKAYSLISpecifications
AAH00019.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
100.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KCAD | |
CDH6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
1004 | |
CDH6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRTYRYFLLLFWVGQPYPTLSTPLSKRTSGFPAKKRALELSGNSKNELNRSKRSWMWNQFFLLEEYTGSDYQYVGKLHSDQDRGDGSLKYILSGDGAGDLFIINENTGDIQATKRLDREEKPVYILRAQAINRRTGRPVEPESEFIIKIHDINDNEPIFTKEVYTATVPEMSDVGTFVVQVTATDADDPTYGNSAKVVYSILQGQPYFSVESETGIIKTALLNMDRENREQYQVVIQAKDMGGQMGGLSGTTTVNITLTDVNDNPPRFPQSTYQFKTPESSPPGTPIGRIKASDADVGENAEIEYSITDGEGLDMFDVITDQETQEGIITVKKLLDFEKKKVYTLKVEASNPYVEPRFLYLGPFKDSATVRIVVEDVDEPPVFSKLAYILQIREDAQINTTIGSVTAQDPDAARNPVKYSVDRHTDMDRIFNIDSGNGSIFTSKLLDRETLLWHNITVIATEINNPKQSSRVPLYIKVLDVNDNAPEFAEFYETFVCEKAKADQLIQTLHAVDKDDPYSGHQFSFSLAPEAASGSNFTIQDNKDNTAGILTRKNGYNRHEMSTYLLPVVISDNDYPVQSSTGTVTVRVCACDHHGNMQSCHAEALIHPTGLSTGALVAILLCIVILLGKLVLPASYLPMVRGSHCYCDTLDLSASPIKAYSLI | |
RUO | |
CDH6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |