missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CDH26 Full-length ORF (NP_068582.2, 1 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00060437-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193203

  • 349.00€ / 10µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

Cadherins are a family of adhesion molecules that mediate Ca2+-dependent cell-cell adhesion in all solid tissues and modulate a wide variety of processes, including cell polarization and migration. Cadherin domains occur as repeats in the extracellular region and are thought to contribute to the sorting of heterogeneous cell types and the maintenance of orderly structures such as epithelium. This gene encodes a cadherin domain-containing protein whose specific function has not yet been determined. Alternative splicing occurs at this locus and two transcript variants, encoding distinct proteins, have been identified. [provided by RefSeq]

Sequence: MKPLIWTWSDVEGQRPALLICTAAAGPTQGVKDLEEVPPSAASQSAQARCALGSWGYGKPFEPRSVKNIHSTPAYPDATMHRQLLAPVEGRMAETLNQKLHVANVLEDDPGYLPHVYSEEGECGGAPSLSSLASLEQELQPDLLDSLGSKATPFEEIYSESGVPS
Specifications

Specifications

NP_068582.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
VR20
CDH26
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
60437
CDH26 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKPLIWTWSDVEGQRPALLICTAAAGPTQGVKDLEEVPPSAASQSAQARCALGSWGYGKPFEPRSVKNIHSTPAYPDATMHRQLLAPVEGRMAETLNQKLHVANVLEDDPGYLPHVYSEEGECGGAPSLSSLASLEQELQPDLLDSLGSKATPFEEIYSESGVPS
RUO
CDH26
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers