Learn More
Abnova™ Human CDC40 Full-length ORF (NP_056975.1, 1 a.a. - 579 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051362-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is found to be essential for the catalytic step II in pre-mRNA splicing process. It is found in the spliceosome, and contains seven WD repeats, which function in protein-protein interactions. This protein has a sequence similarity to yeast Prp17 protein, which functions in two different cellular processes: pre-mRNA splicing and cell cycle progression. It suggests that this protein may play a role in cell cycle progression. [provided by RefSeq]
Sequence: MSAAIAALAASYGSGSGSESDSDSESSRCPLPAADSLMHLTKSPSSKPSLAVAVDSAPEVAVKEDLETGVHLDPAVKEVQYNPTYETMFAPEFGPENPFRTQQMAAPRNMLSGYAEPAHINDFMFEQQRRTFATYGYALDPSLDNHQVSAKYIGSVEEAEKNQGLTVFETGQKKTEKRKKFKENDASNIDGFLGPWAKYVDEKDVAKPSEEEQKELDEITAKRQKKGKQEEEKPGEEKTILHVKEMYDYQGRSYLHIPQDVGVNLRSTMPPEKCYLPKKQIHVWSGHTKGVSAVRLFPLSGHLLLSCSMDCKIKLWEVYGERRCLRTFIGHSKAVRDICFNTAGTQFLSAAYDRYLKLWDTETGQCISRFTNRKVPYCVKFNPDEDKQNLFVAGMSDKKIVQWDIRSGEIVQEYDRHLGAVNTIVFVDENRRFVSTSDDKSLRVWEWDIPVDFKYIAEPSMHSMPAVTLSPNGKWLACQSMDNQILIFGAQNRFRLNKKKIFKGHMVAGYACQVDFSPDMSYVISGDGNGKLNIWDWKTTKLYSRFKAHDKVCIGAVWHPHETSKVITCGWDGLIKLWDSpecifications
NP_056975.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EHB3/FLJ10564/MGC102802/PRP17/PRPF17 | |
CDC40 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51362 | |
CDC40 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSAAIAALAASYGSGSGSESDSDSESSRCPLPAADSLMHLTKSPSSKPSLAVAVDSAPEVAVKEDLETGVHLDPAVKEVQYNPTYETMFAPEFGPENPFRTQQMAAPRNMLSGYAEPAHINDFMFEQQRRTFATYGYALDPSLDNHQVSAKYIGSVEEAEKNQGLTVFETGQKKTEKRKKFKENDASNIDGFLGPWAKYVDEKDVAKPSEEEQKELDEITAKRQKKGKQEEEKPGEEKTILHVKEMYDYQGRSYLHIPQDVGVNLRSTMPPEKCYLPKKQIHVWSGHTKGVSAVRLFPLSGHLLLSCSMDCKIKLWEVYGERRCLRTFIGHSKAVRDICFNTAGTQFLSAAYDRYLKLWDTETGQCISRFTNRKVPYCVKFNPDEDKQNLFVAGMSDKKIVQWDIRSGEIVQEYDRHLGAVNTIVFVDENRRFVSTSDDKSLRVWEWDIPVDFKYIAEPSMHSMPAVTLSPNGKWLACQSMDNQILIFGAQNRFRLNKKKIFKGHMVAGYACQVDFSPDMSYVISGDGNGKLNIWDWKTTKLYSRFKAHDKVCIGAVWHPHETSKVITCGWDGLIKLWD | |
RUO | |
CDC40 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |