Learn More
Abnova™ Human CDC2L2 Partial ORF (NP_277073, 681 a.a. - 780 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00728642-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the p34Cdc2 protein kinase family. p34Cdc2 kinase family members are known to be essential for eukaryotic cell cycle control. This gene is in close proximity to CDC2L1, a nearly identical gene in the same chromosomal region. The gene loci including this gene, CDC2L1, as well as metalloprotease MMP21/22, consist of two identical, tandemly linked genomic regions, which are thought to be a part of the larger region that has been duplicated. This gene and CDC2L1 were shown to be deleted or altered frequently in neuroblastoma with amplified MYCN genes. The protein kinase encoded by this gene could be cleaved by caspases and was demonstrated to play roles in cell apoptosis. Many transcript variants encoding several different isoforms have been found for this gene, but the full-length nature of only two have been determined so far. [provided by RefSeq]
Sequence: GFDLMNKFLTYFPGRRISAEDGLKHEYFRETPLPIDPSMFPTWPAKSEQQRVKRGTSPRPPEGGLGYSQLGDDDLKETGFHLTTTNQGASAAGPGFSLKFSpecifications
NP_277073 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GFDLMNKFLTYFPGRRISAEDGLKHEYFRETPLPIDPSMFPTWPAKSEQQRVKRGTSPRPPEGGLGYSQLGDDDLKETGFHLTTTNQGASAAGPGFSLKF | |
RUO | |
CDC2L2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
728642 | |
CDC2L2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDC2L3/CDK11-p110/CDK11-p46/CDK11-p58/MGC131975/PITSLRE/p58GTA | |
CDC2L2 | |
Recombinant | |
wheat germ expression system |