Learn More
Abnova™ Human CD97 Partial ORF (NP_510966, 421 a.a. - 529 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000976-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the EGF-TM7 family of class II seven-span transmembrane (7-TM) molecules, likely encoded by a gene cluster on the short arm of chromosome 19. The encoded product is a glycoprotein that is present on the surface of most activated leukocytes and spans the membrane seven times, which is a defining feature of G protein-coupled receptors. The protein has an extended extracellular region with several N-terminal epidermal growth factor (EGF)-like domains, which mediate binding to its cellular ligand, decay accelerating factor (DAF, CD55), a regulatory protein of the complement cascade. The presence of structural features characteristic of extracellular matrix proteins and transmembrane proteins suggests that this protein is a receptor involved in both cell adhesion and signaling processes early after leukocyte activation. Alternative splicing has been observed for this gene and three variants have been found. [provided by RefSeq]
Sequence: KKQAELEEIYESSIRGVQLRRLSAVNSIFLSHNNTKELNSPILFAFSHLESSDGEAGRDPPAKDVMPGPRQELLCAFWKSDSDRGGHWATEGCQVLGSKNGSTTCQCSHSpecifications
NP_510966 | |
Liquid | |
976 | |
CD97 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TM7LN1 | |
CD97 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KKQAELEEIYESSIRGVQLRRLSAVNSIFLSHNNTKELNSPILFAFSHLESSDGEAGRDPPAKDVMPGPRQELLCAFWKSDSDRGGHWATEGCQVLGSKNGSTTCQCSH | |
RUO | |
CD97 | |
Wheat Germ (in vitro) | |
GST |