missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CD5 Partial ORF (ABM83003, 403 a.a. - 495 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000921-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: KKLVKKFRQKKQRQWIGPTGMNQNMSFHRNHTATVRSHAENPTASHVDNEYSQPPRNSRLSAYPALEGVLHRSSMQPDNSSDSDYDLHGAQRLSpecifications
ABM83003 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.86kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
KKLVKKFRQKKQRQWIGPTGMNQNMSFHRNHTATVRSHAENPTASHVDNEYSQPPRNSRLSAYPALEGVLHRSSMQPDNSSDSDYDLHGAQRL | |
RUO | |
CD5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
921 | |
CD5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LEU1/T1 | |
CD5 | |
Recombinant | |
wheat germ expression system |