Learn More
ACROBiosystems Human CD40 Ligand / TNFSF5 Protein, Fc tag
Brand: ACROBiosystems CDL-H5267-100UG
Additional Details : Weight : 0.00001kg
Description
CD40 ligand is also known as CD40L, CD154, TNFSF5 and T-cell antigen Gp39, is a single-pass type I I membrane protein which belongs to the TNF superfamily of molecules. CD40 ligand is expressed predominantly on activated CD4+ T lymphocytes, and also found in other types of cells, including platelets, mast cells, macrophages, basophils, NK cells, B lymphocytes, as well as non-haematopoietic cells (smooth muscle cells, endothelial cells, and epithelial cells). Although all monomeric, dimeric and trimeric forms of soluble CD40 ligand can bind to CD40, the trimeric form of soluble CD40 ligand has the most potent biological activity through oligomerization of cell surface CD40, a common feature of TNF receptor family members.CD40 ligand binds to CD40 on antigen-presenting cells (APC), which leads to many effects depending on the target cell type. In general, CD40 ligand plays the role of a costimulatory molecule and induces activation in APC in association with T cell receptor stimulation by MHC molecules on the APC. In total CD40 ligand has three binding partners: CD40, α5β1 integrin and αIIbβ3. CD40 ligand regulates B cell function by engaging CD40 on the B cell surface. A defect in this gene results in an inability to undergo immunoglobulin class switch and is associated with hyper IgM syndrome.
This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 43.3 kDa. As a result of glycosylation, the protein migrates as 45-50 kDa under reducing (R) condition, and 100-115 kDa under non-reducing (NR) condition (SDS-PSpecifications
NP_000065.1 | |
100 μg | |
Met 113 - Leu 261 | |
CD40LG,CD154,CD40L,HIGM1,IGM,IMD3,T-BAM,TNFSF5,TRAP,gp39 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
43.3 kDa | |
Human CD40 Ligand, Fc Tag (CDL-H5267) is expressed from human 293 cells (HEK293). It contains AA Met 113 - Leu 261 (Accession # P29965-1). | |
-20°C | |
CD40 Ligand | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |