missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human CD36 / SR-B3 Protein, His Tag
Brand: ACROBiosystems CD6-H5221-100UG
Additional Details : Weight : 0.00001kg
Description
CD36 (Cluster of Differentiation 36) is also known as platelet membrane glycoprotein IV (GPIV), fatty acid translocase (FAT), thrombospondin receptor, collagen receptor, and scavenger receptor class B, member 3 (SRB3), is a member of the class B scavenger receptor family of cell surface proteins. The human CD36 gene encodes a single chain 472 amino acid residue protein containing both an N- and a C-terminal cytoplasmic tail and an extracellular loop.CD36 is found on platelets, erythrocytes, monocytes, differentiated adipocytes, mammary epithelial cells, spleen cells and some skin microdermal endothelial cells. CD36 is a multiligand pattern recognition receptor that interacts with a large number of structurally dissimilar ligands, including long chain fatty acid (LCFA), advanced glycation end products (AGE), thrombospondin-1, oxidized low-density lipoproteins (oxLDLs), high density lipoprotein (HDL), phosphatidylserine, apoptotic cells, beta-amyloid fibrils (fAβ), collagens I and IV, and Plasmodium falciparum infected erythrocytes. CD36 is required for the anti-angiogenic effects of thrombospondin1 In the corneal neovascularization assay. On binding a ligand the protein and ligand are internalized. This internalization is independent of macropinocytosis and occurs by an actin dependent mechanism requiring the activation Src-family kinases, JNK and Rho-family GTPases. CD36 ligands have also been shown to promote sterile inflammation through assembly of a Toll-like receptor 4 and 6 heterodimer. This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 48.5 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_001001547.1 | |
100 μg | |
Gly 30 - Asn 439 | |
CD36,SCARB3,BDPLT10,CHDS7,FAT,GP3B,GP4,GPIV,PASIV,Platelet Glycoprotein 4,glycoprotein IV,gpIV,glycoprotein IIIb,gpIIIb | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
48.5 kDa | |
Human CD36, His Tag (CD6-H5221) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Asn 439 (Accession # NP_001001547.1). | |
-20°C | |
CD36 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |