Learn More
Abnova™ Human CD300C Partial ORF (NP_006669, 21 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010871-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The CMRF35 antigen, which was identified by reactivity with a monoclonal antibody, is present on monocytes, neutrophils, and some T and B lymphocytes (Jackson et al., 1992 [PubMed 1349532]).[supplied by OMIM]
Sequence: GYFPLSHPMTVAGPVGGSLSVQCRYEKEHRTLNKFWCRPPQILRCDKIVETKGSAGKRNGRVSIRDSPANLSFTVTLENLTEEDAGTYWCGVDTPWLRDFHDPIVEVEVSSpecifications
NP_006669 | |
Liquid | |
10871 | |
CD300C (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CMRF-35A/CMRF35/CMRF35A/CMRF35A1/IGSF16/LIR | |
CD300C | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GYFPLSHPMTVAGPVGGSLSVQCRYEKEHRTLNKFWCRPPQILRCDKIVETKGSAGKRNGRVSIRDSPANLSFTVTLENLTEEDAGTYWCGVDTPWLRDFHDPIVEVEVS | |
RUO | |
CD300C | |
Wheat Germ (in vitro) | |
GST |