missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CD248 Full-length ORF (NP_065137.1, 18 a.a. - 757 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00057124-P03.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102513

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

Sequence: QDPWAAEPRAACGPSSCYALFPRRRTFLEAWRACRELGGDLATPRTPEEAQRVDSLVGAGPASRLLWIGLQRQARQCQLQRPLRGFTWTTGDQDTAFTNWAQPASGGPCPAQRCVALEASGEHRWLEGSCTLAVDGYLCQFGFEGACPALQDEAGQAGPAVYTTPFHLVSTEFEWLPFGSVAAVQCQAGRGASLLCVKQPEGGVGWSRAGPLCLGTGCSPDNGGCEHECVEEVDGHVSCRCTEGFRLAADGRSCEDPCAQAPCEQQCEPGGPQGYSCHCRLGFRPAEDDPHRCVDTDECQIAGVCQQMCVNYVGGFECYCSEGHELEADGISCSPAGAMGAQASQDLGDELLDDGEDEEDEDEAWKAFNGGWTEMPGILWMEPTQPPDFALAYRPSFPEDREPQIPYPEPTWPPPLSAPRVPYHSSVLSVTRPVVVSATHPTLPSAHQPPVIPATHPALSRDHQIPVIAANYPDLPSAYQPGILSVSHSAQPPAHQPPMISTKYPELFPAHQSPMFPDTRVAGTQTTTHLPGIPPNHAPLVTTLGAQLPPQAPDALVLRTQATQLPIIPTAQPSLTTTSRSPVSPAHQISVPAATQPAALPTLLPSQSPTNQTSPISPTHPHSKAPQIPREDGPSPKLALWLPSPAPTAAPTALGEAGLAEHSQRDDRWLLVALLVPTCVFLVVLLALGIVYCTRCGPHAPNKRITDCYRWVIHAGSKSPTEPMPPRGSLTGVQTCRTSV
Specifications

Specifications

NP_065137.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
106.81kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD164L1/MGC119478/MGC119479/TEM1
CD248
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
57124
CD248 (Human) Recombinant Protein (P03)
12.5% SDS-PAGE Stained with Coomassie Blue.
QDPWAAEPRAACGPSSCYALFPRRRTFLEAWRACRELGGDLATPRTPEEAQRVDSLVGAGPASRLLWIGLQRQARQCQLQRPLRGFTWTTGDQDTAFTNWAQPASGGPCPAQRCVALEASGEHRWLEGSCTLAVDGYLCQFGFEGACPALQDEAGQAGPAVYTTPFHLVSTEFEWLPFGSVAAVQCQAGRGASLLCVKQPEGGVGWSRAGPLCLGTGCSPDNGGCEHECVEEVDGHVSCRCTEGFRLAADGRSCEDPCAQAPCEQQCEPGGPQGYSCHCRLGFRPAEDDPHRCVDTDECQIAGVCQQMCVNYVGGFECYCSEGHELEADGISCSPAGAMGAQASQDLGDELLDDGEDEEDEDEAWKAFNGGWTEMPGILWMEPTQPPDFALAYRPSFPEDREPQIPYPEPTWPPPLSAPRVPYHSSVLSVTRPVVVSATHPTLPSAHQPPVIPATHPALSRDHQIPVIAANYPDLPSAYQPGILSVSHSAQPPAHQPPMISTKYPELFPAHQSPMFPDTRVAGTQTTTHLPGIPPNHAPLVTTLGAQLPPQAPDALVLRTQATQLPIIPTAQPSLTTTSRSPVSPAHQISVPAATQPAALPTLLPSQSPTNQTSPISPTHPHSKAPQIPREDGPSPKLALWLPSPAPTAAPTALGEAGLAEHSQRDDRWLLVALLVPTCVFLVVLLALGIVYCTRCGPHAPNKRITDCYRWVIHAGSKSPTEPMPPRGSLTGVQTCRTSV
RUO
CD248
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers