missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CCNH Full-length ORF (NP_001230.1, 1 a.a. - 323 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000902-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16196077

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin forms a complex with CDK7 kinase and ring finger protein MAT1. The kinase complex is able to phosphorylate CDK2 and CDC2 kinases, thus functions as a CDK-activating kinase (CAK). This cyclin and its kinase partner are components of TFIIH, as well as RNA polymerase II protein complexes. They participate in two different transcriptional regulation processes, suggesting an important link between basal transcription control and the cell cycle machinery. [provided by RefSeq]

Sequence: MYHNSSQKRHWTFSSEEQLARLRADANRKFRCKAVANGKVLPNDPVFLEPHEEMTLCKYYEKRLLEFCSVFKPAMPRSVVGTACMYFKRFYLNNSVMEYHPRIIMLTCAFLACKVDEFNVSSPQFVGNLRESPLGQEKALEQILEYELLLIQQLNFHLIVHNPYRPFEGFLIDLKTRYPILENPEILRKTADDFLNRIALTDAYLLYTPSQIALTAILSSASRAGITMESYLSESLMLKENRTCLSQLLDIMKSMRNLVKKYEPPRSEEVAVLKQKLERCHSAELALNVITKKRKGYEDDDYVSKKSKHEEEEWTDDDLVESL
Specifications

Specifications

NP_001230.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAK/p34/p37
CCNH
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
902
CCNH (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MYHNSSQKRHWTFSSEEQLARLRADANRKFRCKAVANGKVLPNDPVFLEPHEEMTLCKYYEKRLLEFCSVFKPAMPRSVVGTACMYFKRFYLNNSVMEYHPRIIMLTCAFLACKVDEFNVSSPQFVGNLRESPLGQEKALEQILEYELLLIQQLNFHLIVHNPYRPFEGFLIDLKTRYPILENPEILRKTADDFLNRIALTDAYLLYTPSQIALTAILSSASRAGITMESYLSESLMLKENRTCLSQLLDIMKSMRNLVKKYEPPRSEEVAVLKQKLERCHSAELALNVITKKRKGYEDDDYVSKKSKHEEEEWTDDDLVESL
RUO
CCNH
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers