missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CCNF Full-length ORF (NP_001752.2, 1 a.a. - 786 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000899-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16156077

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the cyclin family. Cyclins are important regulators of cell cycle transitions through their ability to bind and activate cyclin-dependent protein kinases. This member also belongs to the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it was one of the first proteins in which the F-box motif was identified. [provided by RefSeq]

Sequence: MGSGGVVHCRCAKCFCYPTKRRIRRRPRNLTILSLPEDVLFHILKWLSVEDILAVRAVHSQLKDLVDNHASVWACASFQELWPSPGNLKLFERAAEKGNFEAAVKLGIAYLYNEGLSVSDEARAEVNGLKASRFFSLAERLNVGAAPFIWLFIRPPWSVSGSCCKAVVHESLRAECQLQRTHKASILHCLGRVLSLFEDEEKQQQAHDLFEEAAHQGCLTSSYLLWESDRRTDVSDPGRCLHSFRKLRDYAAKGCWEAQLSLAKACANANQLGLEVRASSEIVCQLFQASQAVSKQQVFSVQKGLNDTMRYILIDWLVEVATMKDFTSLCLHLTVECVDRYLRRRLVPRYRLQLLGIACMVICTRFISKEILTIREAVWLTDNTYKYEDLVRMMGEIVSALEGKIRVPTVVDYKEVLLTLVPVELRTQHLCSFLCELSLLHTSLSAYAPARLAAAALLLARLTHGQTQPWTTQLWDLTGFSYEDLIPCVLSLHKKCFHDDAPKDYRQVSLTAVKQRFEDKRYGEISQEEVLSYSQLCAALGVTQDSPDPPTFLSTGEIHAFLSSPSGRRTKRKRENSLQEDRGSFVTTPTAELSSQEETLLGSFLDWSLDCCSGYEGDQESEGEKEGDVTAPSGILDVTVVYLNPEQHCCQESSDEEACPEDKGPQDPQALALDTQIPATPGPKPLVRTSREPGKDVTTSGYSSVSTASPTSSVDGGLGALPQPTSVLSLDSDSHTQPCHHQARKSCLQCRPPSPPESSVPQQQVKRINLCIHSEEEDMNLGLVRL
Specifications

Specifications

NP_001752.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
114kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FBX1/FBXO1
CCNF
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
899
CCNF (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGSGGVVHCRCAKCFCYPTKRRIRRRPRNLTILSLPEDVLFHILKWLSVEDILAVRAVHSQLKDLVDNHASVWACASFQELWPSPGNLKLFERAAEKGNFEAAVKLGIAYLYNEGLSVSDEARAEVNGLKASRFFSLAERLNVGAAPFIWLFIRPPWSVSGSCCKAVVHESLRAECQLQRTHKASILHCLGRVLSLFEDEEKQQQAHDLFEEAAHQGCLTSSYLLWESDRRTDVSDPGRCLHSFRKLRDYAAKGCWEAQLSLAKACANANQLGLEVRASSEIVCQLFQASQAVSKQQVFSVQKGLNDTMRYILIDWLVEVATMKDFTSLCLHLTVECVDRYLRRRLVPRYRLQLLGIACMVICTRFISKEILTIREAVWLTDNTYKYEDLVRMMGEIVSALEGKIRVPTVVDYKEVLLTLVPVELRTQHLCSFLCELSLLHTSLSAYAPARLAAAALLLARLTHGQTQPWTTQLWDLTGFSYEDLIPCVLSLHKKCFHDDAPKDYRQVSLTAVKQRFEDKRYGEISQEEVLSYSQLCAALGVTQDSPDPPTFLSTGEIHAFLSSPSGRRTKRKRENSLQEDRGSFVTTPTAELSSQEETLLGSFLDWSLDCCSGYEGDQESEGEKEGDVTAPSGILDVTVVYLNPEQHCCQESSDEEACPEDKGPQDPQALALDTQIPATPGPKPLVRTSREPGKDVTTSGYSSVSTASPTSSVDGGLGALPQPTSVLSLDSDSHTQPCHHQARKSCLQCRPPSPPESSVPQQQVKRINLCIHSEEEDMNLGLVRL
RUO
CCNF
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers