Learn More
Abnova™ Human CCNB3 Partial ORF (NP_149020, 1296 a.a. - 1395 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00085417-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. Studies of similar genes in chick and Drosophila suggest that this cyclin may associate with CDC2 and CDK2 kinases, and be required for proper spindle reorganization and restoration of the interphase nucleus. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: VQEKASKLAAASLLLALYMKKLGYWVPFLEHYSGYSISELHPLVRQLNKLLTFSSYDSLKAVYYKYSHPVFFEVAKIPALDMLKLEEILNCDCEAQGLVLSpecifications
NP_149020 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VQEKASKLAAASLLLALYMKKLGYWVPFLEHYSGYSISELHPLVRQLNKLLTFSSYDSLKAVYYKYSHPVFFEVAKIPALDMLKLEEILNCDCEAQGLVL | |
RUO | |
CCNB3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
85417 | |
CCNB3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCNB3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |