missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CCL4L1 Full-length ORF (AAI48785.1, 1 a.a. - 92 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009560-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16103842

  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. This protein is similar to CCL4 which inhibits HIV entry by binding to the cellular receptor CCR5. The copy number of this gene varies among individuals; most individuals have 1-5 copies in the diploid genome, although rare individuals do not contain this gene at all. The human genome reference assembly contains two copies of this gene. This record represents the more centromeric gene. [provided by RefSeq]

Sequence: MKLCVTVLSLLVLVAAFCSLALSAPMGSDPPTACCFSYTARKLPRNFVVDYYETSSLCSQPAVVFQTKRGKQVCADPSESWVQEYVYDLELN
Specifications

Specifications

AAI48785.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.07kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AT744.2/CCL4L/LAG-1/LAG1/SCYA4L
CCL4L1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9560
CCL4L1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKLCVTVLSLLVLVAAFCSLALSAPMGSDPPTACCFSYTARKLPRNFVVDYYETSSLCSQPAVVFQTKRGKQVCADPSESWVQEYVYDLELN
RUO
CCL4L1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers