missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CCL3 Partial ORF (NP_002974, 24 a.a. - 92 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006348-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105015

  • 349.00€ / 10µg

Estimated Shipment: 07-07-2023
to see stock.
Add to basket

Description

Description

Macrophage inflammatory protein-1 is a so-called monokine that is involved in the acute inflammatory state in the recruitment and activation of polymorphonuclear leukocytes (Wolpe et al., 1988 [PubMed 3279154]). Sherry et al. (1988) [PubMed 3058856] demonstrated 2 protein components of MIP1, called by them alpha and beta.[supplied by OMIM]

Sequence: SLAADTPTACCFSYTSRQIPQNFIADYFETSSQCSKPGVIFLTKRSRQVCADPSEEWVQKYVSDLELSA
Specifications

Specifications

NP_002974
Liquid
6348
CCL3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
G0S19-1/LD78ALPHA/MIP-1-alpha/MIP1A/SCYA3
CCL3
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.33kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SLAADTPTACCFSYTSRQIPQNFIADYFETSSQCSKPGVIFLTKRSRQVCADPSEEWVQKYVSDLELSA
RUO
CCL3
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers