Learn More
Abnova™ Human CBY1 Full-length ORF (NP_001002880.1, 1 a.a. - 126 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00025776-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Beta-catenin is a transcriptional activator and oncoprotein involved in the development of several cancers. The protein encoded by this gene interacts directly with the C-terminal region of beta-catenin, inhibiting oncogenic beta-catenin-mediated transcriptional activation by competing with transcription factors for binding to beta-catenin. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MPFFGNTFSPKKTPPRKSASLSNLHSLDRSTREVELGLEYGSPTMNLAGQSLKFENGQWIAETGVSGGVDRREVQRLRRRNQQLEEENNLLRLKVDILLDMLSESTAESHLMEKELDELRISRKRKSpecifications
NP_001002880.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C22orf2/CBY/HS508I15A/PGEA1/PIGEA-14/PIGEA14/arb1 | |
CBY1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
25776 | |
CBY1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPFFGNTFSPKKTPPRKSASLSNLHSLDRSTREVELGLEYGSPTMNLAGQSLKFENGQWIAETGVSGGVDRREVQRLRRRNQQLEEENNLLRLKVDILLDMLSESTAESHLMEKELDELRISRKRK | |
RUO | |
CBY1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |