Learn More
Abnova™ Human CBLC Full-length ORF (AAH28915, 1 a.a. - 474 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023624-P01.25ug
Additional Details : Weight : 0.00010kg
Description
CBL proteins, such as CBLC, are phosphorylated upon activation of a variety of receptors that signal via protein tyrosine kinases. Through interactions with proteins containing SRC (MIM 190090) homology-2 (SH2) and SH3 domains, CBL proteins modulate downstream cell signaling (Keane et al., 1999 [PubMed 10362357]).[supplied by OMIM]
Sequence: MALAVAPWGRQWEEARALGRAVRMLQRLEEQCVDPRLSVSPPSLRDLLPRTAQLLREVAHSRRAAGGGGPGGPGGSGDFLLIYLANLEAKSRQVAALLPPRGRRSANDELFRAGSRLRRQLAKLAIIFSHMHAELHALFPGGKYCGHMYQLTKAPAHTFWRESCGARCVLPWAEFESLLGTCHPVEPGCTALALRTTIDLTCSGHVSIFEFDVFTRLFQPWPTLLKNWQLLAVNHPGYMAFLTYDEVQERLQACRDKPGSYIFRPSCTRLGQWAIGYVSSDGSILQTIPANKPLSQVLLEGQKDGFYLYPDGKTHNPDLTELGQAEPQQRIHVSEEQLQLYWAMDSTFELCKICAESNKDVKIEPCGHLLCSCCLAAWQHSDSQTCPFCRCEIKGWEAVSIYQFHGQATAEDPGNSSDQEGRELELGQVPLSAPPLPPRPDLPPRKPRNAQPKVRLLKGNSPPAALGPQDPAPASpecifications
AAH28915 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
77.88kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CBL-3/CBL-SL/RNF57 | |
CBLC | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23624 | |
CBLC (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALAVAPWGRQWEEARALGRAVRMLQRLEEQCVDPRLSVSPPSLRDLLPRTAQLLREVAHSRRAAGGGGPGGPGGSGDFLLIYLANLEAKSRQVAALLPPRGRRSANDELFRAGSRLRRQLAKLAIIFSHMHAELHALFPGGKYCGHMYQLTKAPAHTFWRESCGARCVLPWAEFESLLGTCHPVEPGCTALALRTTIDLTCSGHVSIFEFDVFTRLFQPWPTLLKNWQLLAVNHPGYMAFLTYDEVQERLQACRDKPGSYIFRPSCTRLGQWAIGYVSSDGSILQTIPANKPLSQVLLEGQKDGFYLYPDGKTHNPDLTELGQAEPQQRIHVSEEQLQLYWAMDSTFELCKICAESNKDVKIEPCGHLLCSCCLAAWQHSDSQTCPFCRCEIKGWEAVSIYQFHGQATAEDPGNSSDQEGRELELGQVPLSAPPLPPRPDLPPRKPRNAQPKVRLLKGNSPPAALGPQDPAPA | |
RUO | |
CBLC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |