missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CAMK1G Partial ORF (AAH32787, 371 a.a. - 476 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00057172-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192573

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes a protein similar to calcium/calmodulin dependent protein kinase, however, its exact function is not known. [provided by RefSeq]

Sequence: TQLPCQHGRRPTAPGGRSLNCLVNGSLHISSSLVPMHQGSLAAGPCGCCSSCLNIGSKGKSSYCSEPTLLKKANKKQNFKSEVMVPVKASGSSHCRAGQTGVCLIM
Specifications

Specifications

AAH32787
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.29kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TQLPCQHGRRPTAPGGRSLNCLVNGSLHISSSLVPMHQGSLAAGPCGCCSSCLNIGSKGKSSYCSEPTLLKKANKKQNFKSEVMVPVKASGSSHCRAGQTGVCLIM
RUO
CAMK1G
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
57172
CAMK1G (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CLICKIII/VWS1/dJ272L16.1
CAMK1G
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers