missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CALD1 Full-length ORF (NP_004333.1, 1 a.a. - 538 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000800-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195947

  • 349.00€ / 10µg

Estimated Shipment: 10-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes a calmodulin- and actin-binding protein that plays an essential role in the regulation of smooth muscle and nonmuscle contraction. The conserved domain of this protein possesses the binding activities to Ca(2+)-calmodulin, actin, tropomyosin, myosin, and phospholipids. This protein is a potent inhibitor of the actin-tropomyosin activated myosin MgATPase, and serves as a mediating factor for Ca(2+)-dependent inhibition of smooth muscle contraction. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]

Sequence: MDDFERRRELRRQKREEMRLEAERIAYQRNDDDEEEAARERRRRARQERLRQKQEEESLGQVTDQVEVNAQNSVPDEEAKTTTTNTQVEGDDEAAFLERLARREERRQKRLQEALERQKEFDPTITDASLSLPSRRMQNDTAENETTEKEEKSESRQERYEIEETETVTKSYQKNDWRDAEENKKEDKEKEEEEEEKPKRGSIGENQIKDEKIKKDKEPKEEVKSFMDRKKGFTEVKSQNGEFMTHKLKHTENTFSRPGGRASVDTKEAEGAPQVEAGKRLEELRRRRGETESEEFEKLKQKQQEAALELEELKKKREERRKVLEEEEQRRKQEEADRKLREEEEKRRLKEEIERRRAEAAEKRQKMPEDGLSDDKKPFKCFTPKGSSLKIEERAEFLNKSVQKSSGVKSTHQAAIVSKIDSRLEQYTSAIEGTKSAKPTKPAASDLPVPAEGVRNIKSMWEKGNVFSSPTAAGTPNKETAGLKVGVSSRINEWLTKTPDGNKSPAPKPSDLRPGDVSSKRNLWEKQSVDKVTSPTKV
Specifications

Specifications

NP_004333.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
89.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CDM/H-CAD/L-CAD/MGC21352/NAG22
CALD1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
800
CALD1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDDFERRRELRRQKREEMRLEAERIAYQRNDDDEEEAARERRRRARQERLRQKQEEESLGQVTDQVEVNAQNSVPDEEAKTTTTNTQVEGDDEAAFLERLARREERRQKRLQEALERQKEFDPTITDASLSLPSRRMQNDTAENETTEKEEKSESRQERYEIEETETVTKSYQKNDWRDAEENKKEDKEKEEEEEEKPKRGSIGENQIKDEKIKKDKEPKEEVKSFMDRKKGFTEVKSQNGEFMTHKLKHTENTFSRPGGRASVDTKEAEGAPQVEAGKRLEELRRRRGETESEEFEKLKQKQQEAALELEELKKKREERRKVLEEEEQRRKQEEADRKLREEEEKRRLKEEIERRRAEAAEKRQKMPEDGLSDDKKPFKCFTPKGSSLKIEERAEFLNKSVQKSSGVKSTHQAAIVSKIDSRLEQYTSAIEGTKSAKPTKPAASDLPVPAEGVRNIKSMWEKGNVFSSPTAAGTPNKETAGLKVGVSSRINEWLTKTPDGNKSPAPKPSDLRPGDVSSKRNLWEKQSVDKVTSPTKV
RUO
CALD1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers