Learn More
Abnova™ Human CALD1 Full-length ORF (NP_004333.1, 1 a.a. - 538 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000800-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a calmodulin- and actin-binding protein that plays an essential role in the regulation of smooth muscle and nonmuscle contraction. The conserved domain of this protein possesses the binding activities to Ca(2+)-calmodulin, actin, tropomyosin, myosin, and phospholipids. This protein is a potent inhibitor of the actin-tropomyosin activated myosin MgATPase, and serves as a mediating factor for Ca(2+)-dependent inhibition of smooth muscle contraction. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]
Sequence: MDDFERRRELRRQKREEMRLEAERIAYQRNDDDEEEAARERRRRARQERLRQKQEEESLGQVTDQVEVNAQNSVPDEEAKTTTTNTQVEGDDEAAFLERLARREERRQKRLQEALERQKEFDPTITDASLSLPSRRMQNDTAENETTEKEEKSESRQERYEIEETETVTKSYQKNDWRDAEENKKEDKEKEEEEEEKPKRGSIGENQIKDEKIKKDKEPKEEVKSFMDRKKGFTEVKSQNGEFMTHKLKHTENTFSRPGGRASVDTKEAEGAPQVEAGKRLEELRRRRGETESEEFEKLKQKQQEAALELEELKKKREERRKVLEEEEQRRKQEEADRKLREEEEKRRLKEEIERRRAEAAEKRQKMPEDGLSDDKKPFKCFTPKGSSLKIEERAEFLNKSVQKSSGVKSTHQAAIVSKIDSRLEQYTSAIEGTKSAKPTKPAASDLPVPAEGVRNIKSMWEKGNVFSSPTAAGTPNKETAGLKVGVSSRINEWLTKTPDGNKSPAPKPSDLRPGDVSSKRNLWEKQSVDKVTSPTKVSpecifications
NP_004333.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
89.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDM/H-CAD/L-CAD/MGC21352/NAG22 | |
CALD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
800 | |
CALD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDDFERRRELRRQKREEMRLEAERIAYQRNDDDEEEAARERRRRARQERLRQKQEEESLGQVTDQVEVNAQNSVPDEEAKTTTTNTQVEGDDEAAFLERLARREERRQKRLQEALERQKEFDPTITDASLSLPSRRMQNDTAENETTEKEEKSESRQERYEIEETETVTKSYQKNDWRDAEENKKEDKEKEEEEEEKPKRGSIGENQIKDEKIKKDKEPKEEVKSFMDRKKGFTEVKSQNGEFMTHKLKHTENTFSRPGGRASVDTKEAEGAPQVEAGKRLEELRRRRGETESEEFEKLKQKQQEAALELEELKKKREERRKVLEEEEQRRKQEEADRKLREEEEKRRLKEEIERRRAEAAEKRQKMPEDGLSDDKKPFKCFTPKGSSLKIEERAEFLNKSVQKSSGVKSTHQAAIVSKIDSRLEQYTSAIEGTKSAKPTKPAASDLPVPAEGVRNIKSMWEKGNVFSSPTAAGTPNKETAGLKVGVSSRINEWLTKTPDGNKSPAPKPSDLRPGDVSSKRNLWEKQSVDKVTSPTKV | |
RUO | |
CALD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |