missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CACNA1E Partial ORF (NP_000712, 1901 a.a. - 1999 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000777-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105504

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes an alpha-1 subunit of a voltage-dependent calcium channel. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization. The alpha-1 subunit consists of 24 transmembrane segments and forms the pore through which ions pass into the cell. The calcium channel consists of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. There are multiple isoforms of each of these proteins, either encoded by different genes or the result of alternative splicing of transcripts. Alternate transcriptional splice variants of the gene described here have been observed but have not been thoroughly characterized. [provided by RefSeq]

Sequence: RMEPSSLPQEIIANAKALPYLQQDPVSGLSGRSGYPSMSPLSPQDIFQLACMDPADDGQFQERQSLVVTDPSSMRRSFSTIRDKRSNSSWLEEFSMERS
Specifications

Specifications

NP_000712
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RMEPSSLPQEIIANAKALPYLQQDPVSGLSGRSGYPSMSPLSPQDIFQLACMDPADDGQFQERQSLVVTDPSSMRRSFSTIRDKRSNSSWLEEFSMERS
RUO
CACNA1E
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
777
CACNA1E (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BII/CACH6/CACNL1A6/Cav2.3
CACNA1E
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers