missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C9orf4 Partial ORF (NP_055149, 158 a.a. - 267 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023732-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105016

  • 508.00€ / 25µg
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

Sequence: FRYGKPGCNAETCDYFLSYRMIGADVEFELSADTDGWVAVGFSSDKKMGGDDVMACVHDDNGRVRIQHFYNVGQWAKEIQRNPARDEEGVFENNRVTCRFKRPVNVPRDE
Specifications

Specifications

NP_055149
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FRYGKPGCNAETCDYFLSYRMIGADVEFELSADTDGWVAVGFSSDKKMGGDDVMACVHDDNGRVRIQHFYNVGQWAKEIQRNPARDEEGVFENNRVTCRFKRPVNVPRDE
RUO
C9orf4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
23732
C9orf4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CG-6/CG6
C9orf4
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers