missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C9orf23 Full-length ORF (NP_680544.1, 1 a.a. - 163 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00138716-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16128133

  • 508.00€ / 25µg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

This gene encodes a protein that appears to belong to a family of evolutionarily related proteins (DUF78), that may share one or more domains in common. Members of this family are small archaebacterial proteins with no known function. Alternative splicing has been observed at this locus and two variants, both encoding the same protein, have been identified. [provided by RefSeq]

Sequence: MEHYRKAGSVELPAPSPMPQLPPDTLEMRVRDGSKIRNLLGLALGRLEGGSARHVVFSGSGRAAGKAVSCAEIVKRRVPGLHQLTKLRFLQTEDSWVPASPDTGLDPLTVRRHVPAVWVLLSRDPLDPNECGYQPPGAPPGLGSMPSSSCGPRSRRRARDTRS
Specifications

Specifications

NP_680544.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.00kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC29635/bA296L22.5
C9orf23
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
138716
C9orf23 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEHYRKAGSVELPAPSPMPQLPPDTLEMRVRDGSKIRNLLGLALGRLEGGSARHVVFSGSGRAAGKAVSCAEIVKRRVPGLHQLTKLRFLQTEDSWVPASPDTGLDPLTVRRHVPAVWVLLSRDPLDPNECGYQPPGAPPGLGSMPSSSCGPRSRRRARDTRS
RUO
C9orf23
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers