missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C17orf38 Partial ORF (NP_001010855, 661 a.a. - 754 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00146850-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192047

  • 528.00€ / 25µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

Phosphoinositide 3-kinase gamma is a lipid kinase that produces the lipid second messenger phosphatidylinositol 3,4,5-trisphosphate. The kinase is composed of a catalytic subunit and one of several regulatory subunits, and is chiefly activated by G protein-coupled receptors. This gene encodes a regulatory subunit, and is distantly related to the phosphoinositide-3-kinase, regulatory subunit 5 gene which is located adjacent to this gene on chromosome 7. The orthologous protein in the mouse binds to both the catalytic subunit and to G(beta/gamma), and mediates activation of the kinase subunit downstream of G protein-coupled receptors. [provided by RefSeq]

Sequence: GKSFSTVTNTFRTNNIQIQSRDQRLLTLSLDKDDQRTFRDVVRFEVAPCPEPCSGAQKSKAPWLNLHGQQEVEAIKAKPKPLLMPINTFSGIVQ
Specifications

Specifications

NP_001010855
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.08kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GKSFSTVTNTFRTNNIQIQSRDQRLLTLSLDKDDQRTFRDVVRFEVAPCPEPCSGAQKSKAPWLNLHGQQEVEAIKAKPKPLLMPINTFSGIVQ
RUO
PIK3R6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
146850
C17orf38 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C17orf38/DKFZp666P158/FLJ34500/HsT41028/p84/p87(PIKAP)/p87PIKAP
PIK3R6
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers