missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C13orf22 Partial ORF (NP_005791, 975 a.a. - 1074 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010208-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: LLSPTPVSTELSENGEGDFRYLGMGDSHIPPPVPSEFNDVSQNTHLRQDHNYCSPTKKNPCEVQPDSLTNNACVRTLNLESPMKTDIFDEFFSSSALNALSpecifications
NP_005791 | |
Liquid | |
10208 | |
C13orf22 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C13orf22/D13S106E/DKFZp781K2286/FLJ32952/RP11-121O19.1/bA121O19.1 | |
USPL1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LLSPTPVSTELSENGEGDFRYLGMGDSHIPPPVPSEFNDVSQNTHLRQDHNYCSPTKKNPCEVQPDSLTNNACVRTLNLESPMKTDIFDEFFSSSALNAL | |
RUO | |
USPL1 | |
Wheat Germ (in vitro) | |
GST |