missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C13orf18 Full-length ORF (AAH43488.1, 1 a.a. - 595 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00080183-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16124813

  • 545.00€ / 25µg

Estimated Shipment: 13-09-2022
to see stock.
Add to basket

Description

Description

Sequence: MVSNHYFLLCVNLPLREIHTPGPSPSCLGDSLAETTLSEDTTDSVGSASPHGSSEKSSSFSLSSTEVHMVRPGYSHRVSLPTSPRILATSPYPETDSAFFEPSHLTSAADEGAVQVSRRTISSNSFSPEVFVLPVDVEKENAHFYVADMIISAMEKMKCNILSQQQTESWSKEVSGLLGSDQPDSEMTFDTNIKQESGSSTSSYSGYEGCAVLQVSPVTETRTYHDVKEICKCDVDEFVILELGDFNDITETCSCSCSSSKSVTYEPDFNSAELLAKELYRVFQKCWILSVVNSQLAGSLSAAGSIVVNEECVRKDFESSMNVVQEIKFKSRIRGTEDWAPPRFQIIFNIHPPLKRDLVVAAQNFFCAGCGTPVEPKFVKRLRYCEYLGKYFCDCCHSYAESCIPARILMMWDFKKYYVSNFSKQLLDSIWHQPIFNLLSIGQSLYAKAKELDRVKEIQEQLFHIKKLLKTCRFANSALKEFEQVPGHLTDELHLFSLEDLVRIKKGLLAPLLKDILKASLAHVAGCELCQGKGFICEFCQNTTVIFPFQTATCRRCSACRACFHKQCFQSSECPRCARITARRKLLESVASAAT
Specifications

Specifications

AAH43488.1
Liquid
80183
C13orf18 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVSNHYFLLCVNLPLREIHTPGPSPSCLGDSLAETTLSEDTTDSVGSASPHGSSEKSSSFSLSSTEVHMVRPGYSHRVSLPTSPRILATSPYPETDSAFFEPSHLTSAADEGAVQVSRRTISSNSFSPEVFVLPVDVEKENAHFYVADMIISAMEKMKCNILSQQQTESWSKEVSGLLGSDQPDSEMTFDTNIKQESGSSTSSYSGYEGCAVLQVSPVTETRTYHDVKEICKCDVDEFVILELGDFNDITETCSCSCSSSKSVTYEPDFNSAELLAKELYRVFQKCWILSVVNSQLAGSLSAAGSIVVNEECVRKDFESSMNVVQEIKFKSRIRGTEDWAPPRFQIIFNIHPPLKRDLVVAAQNFFCAGCGTPVEPKFVKRLRYCEYLGKYFCDCCHSYAESCIPARILMMWDFKKYYVSNFSKQLLDSIWHQPIFNLLSIGQSLYAKAKELDRVKEIQEQLFHIKKLLKTCRFANSALKEFEQVPGHLTDELHLFSLEDLVRIKKGLLAPLLKDILKASLAHVAGCELCQGKGFICEFCQNTTVIFPFQTATCRRCSACRACFHKQCFQSSECPRCARITARRKLLESVASAAT
RUO
C13orf18
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
92.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ21562/FLJ43762
C13orf18
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers