Learn More
R&D Systems™ Human C-Reactive Protein/CRP Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
Unit Size:
Pack of 96
Specifications
Specifications
| Specificity | Natural and recombinant human CRP. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1401 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895 |
| Target | Human |