Learn More
Abnova™ Human BSCL2 Partial ORF (NP_116056, 259 a.a. - 357 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00026580-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes protein seipin, which is located in the endoplasmic reticulum and may be important for lipid droplet morphology. Mutations in this gene have been associated with congenital generalized lipodystrophy type 2 or Berardinelli-Seip syndrome, a rare autosomal recessive disease characterized by a near absence of adipose tissue and severe insulin resistance. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: WGGIWPRHRFSLQVNIRKRDNSRKEVQRRISAHQPGPEGQEESTPQSDVTEDGESPEDPSGTEGQLSEEEKPDQQPLSGEEELEPEASDGSGSWEDAALSpecifications
NP_116056 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
WGGIWPRHRFSLQVNIRKRDNSRKEVQRRISAHQPGPEGQEESTPQSDVTEDGESPEDPSGTEGQLSEEEKPDQQPLSGEEELEPEASDGSGSWEDAAL | |
RUO | |
BSCL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
26580 | |
BSCL2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GNG3LG/HMN5/MGC4694/SPG17 | |
BSCL2 | |
Recombinant | |
wheat germ expression system |