Learn More
Abnova™ Human BRUNOL5 Full-length ORF (AAH47522.1, 1 a.a. - 402 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00060680-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Members of the CELF/BRUNOL protein family contain two N-terminal RNA recognition motif (RRM) domains, one C-terminal RRM domain, and a divergent segment of 160-230 aa between the second and third RRM domains. Members of this protein family regulate pre-mRNA alternative splicing and may also be involved in mRNA editing, and translation. [provided by RefSeq]
Sequence: MARLTESEARRQQQQLLQPRPSPVGSSGPEPPGGQPDGMKDLDAIKLFVGQIPRHLDEKDLKPLLEQFGRIYELTVLKDPYTGMHKGCAFLTYCARDSAIKAQTALHEQKTLPGMARPIQVKPADSESRGGRDRKLFVGMLNKQQSEEDVLRLFQPFGVIDECTVLRGPDGSSKGCAFVKFSSHTEAQAAIHALHGSQTMPGASSSLVVKFADTDKERTLRRMQQMVGQLGILTPSLTLPFSPYSAYAQALMQQQTTVLSTSGSYLSPGVAFSPCHIQQIGAVSLNGLPATPIAPASGLHSPPLLGTTAVPGLVAPITNGFAGVVPFSRWAPCPGNRLCQWPCALPSSEPDCGRDTASCLLRSPAVHSHVPHRGHHAHRAQRPPAAAPPAAAAARRSLETRSSpecifications
AAH47522.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
69.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
BRUNOL5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
60680 | |
BRUNOL5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BRUNOL-5/CELF5 | |
BRUNOL5 | |
Recombinant | |
wheat germ expression system |