missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BRCC3 Full-length ORF (NP_077308.1, 1 a.a. - 316 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079184-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16124153

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a subunit of the BRCA1-BRCA2-containing complex (BRCC), which is an E3 ubiquitin ligase. This protein is also thought to be involved in the cellular response to ionizing radiation and progression through the G2/M checkpoint. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MAVQVVQAVQAVHLESDAFLVCLNHALSTEKEEVMGLCIGELNDDTRSDSKFAYTGTEMRTVAEKVDAVRIVHIHSVIILRRSDKRKDRVEISPEQLSAASTEAERLAELTGRPMRVVGWYHSHPHITVWPSHVDVRTQAMYQMMDQGFVGLIFSCFIEDKNTKTGRVLYTCFQSIQAQKSSESLHGPRDFWSSSQHISIEGQKEEERYERIEIPIHIVPHVTIGKVCLESAVELPKILCQEEQDAYRRIHSLTHLDSVTKIHNGSVFTKNLCSQMSAVSGPLLQWLEDRLEQNQQHLQELQQEKEELMQELSSLE
Specifications

Specifications

NP_077308.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
BRCC3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
79184
BRCC3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BRCC36/C6.1A/CXorf53
BRCC3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers