Learn More
Abnova™ Human BRCA1 Partial ORF (NP_009225, 1581 a.a. - 1670 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_009225 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 672 |
Molecular Weight (g/mol) | 35.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16112341
|
Abnova™
H00000672-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16102341
|
Abnova™
H00000672-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a nuclear phosphoprotein that plays a role in maintaining genomic stability, and it also acts as a tumor suppressor. The encoded protein combines with other tumor suppressors, DNA damage sensors, and signal transducers to form a large multi-subunit protein complex known as the BRCA1-associated genome surveillance complex (BASC). This gene product associates with RNA polymerase II, and through the C-terminal domain, also interacts with histone deacetylase complexes. This protein thus plays a role in transcription, DNA repair of double-stranded breaks, and recombination. Mutations in this gene are responsible for approximately 40% of inherited breast cancers and more than 80% of inherited breast and ovarian cancers. Alternative splicing plays a role in modulating the subcellular localization and physiological function of this gene. Many alternatively spliced transcript variants, some of which are disease-associated mutations, have been described for this gene, but the full-length natures of only some of these variants has been described. A related pseudogene, which is also located on chromosome 17, has been identified. [provided by RefSeq]
Sequence: EDRAPESARVGNIPSSTSALKVPQLKVAESAQGPAAAHTTDTAGYNAMEESVSREKPELTASTERVNKRMSMVVSGLTPEEFMLVYKFARSpecifications
NP_009225 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BRCAI/BRCC1/IRIS/PSCP/RNF53 | |
BRCA1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
672 | |
BRCA1 (Human) Recombinant Protein (Q01) | |
EDRAPESARVGNIPSSTSALKVPQLKVAESAQGPAAAHTTDTAGYNAMEESVSREKPELTASTERVNKRMSMVVSGLTPEEFMLVYKFAR | |
RUO | |
BRCA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |