Learn More
Abnova™ Human BMP8B Partial ORF (NP_001711, 274 a.a. - 362 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000656-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. Based on its expression early in embryogenesis, the BMP encoded by this gene has a proposed role in early development. In addition, the fact that this BMP is closely related to BMP5 and BMP7 has led to speculation of possible bone inductive activity. [provided by RefSeq]
Sequence: KKSNELPQANRLPGIFDDVHGSHGRQVCRRHELYVSFQDLGWLDWVIAPQGYSAYYCEGECSFPLDSCMNATNHAILQSLVHLMMPDAVSpecifications
NP_001711 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KKSNELPQANRLPGIFDDVHGSHGRQVCRRHELYVSFQDLGWLDWVIAPQGYSAYYCEGECSFPLDSCMNATNHAILQSLVHLMMPDAV | |
RUO | |
BMP8B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
656 | |
BMP8B (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BMP8/MGC131757/OP2 | |
BMP8B | |
Recombinant | |
wheat germ expression system |