missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BLMH Full-length ORF (NP_000377.1, 1 a.a. - 455 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_000377.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 642 |
Molecular Weight (g/mol) | 79kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16115757
|
Abnova™
H00000642-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment:
Log in to see stock. |
|||||
16105757
|
Abnova™
H00000642-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment:
Log in to see stock. |
|||||
Description
Bleomycin hydrolase (BMH) is a cytoplasmic cysteine peptidase that is highly conserved through evolution; however, the only known activity of the enzyme is metabolic inactivation of the glycopeptide bleomycin (BLM), an essential component of combination chemotherapy regimens for cancer. The protein contains the signature active site residues of the cysteine protease papain superfamily. [provided by RefSeq]
Sequence: MSSSGLNSEKVAALIQKLNSDPQFVLAQNVGTTHDLLDICLKRATVQRAQHVFQHAVPQEGKPITNQKSSGRCWIFSCLNVMRLPFMKKLNIEEFEFSQSYLFFWDKVERCYFFLSAFVDTAQRKEPEDGRLVQFLLMNPANDGGQWDMLVNIVEKYGVIPKKCFPESYTTEATRRMNDILNHKMREFCIRLRNLVHSGATKGEISATQDVMMEEIFRVVCICLGNPPETFTWEYRDKDKNYQKIGPITPLEFYREHVKPLFNMEDKICLVNDPRPQHKYNKLYTVEYLSNMVGGRKTLYNNQPIDFLKKMVAASIKDGEAVWFGCDVGKHFNSKLGLSDMNLYDHELVFGVSLKNMNKAERLTFGESLMTHAMTFTAVSEKDDQDGAFTKWRVENSWGEDHGHKGYLCMTDEWFSEYVYEVVVDRKHVPEEVLAVLEQEPIILPAWDPMGALAESpecifications
NP_000377.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BH/BMH | |
BLMH | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
642 | |
BLMH (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
BLMH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |