Learn More
R&D Systems™ Human Biomarker Premixed Magnetic Luminex™ Performance Assay Kit A
Product Code. 18779667
Change view
Click to view available options
Quantity:
2 Plex
5 Plex
7 Plex
Unit Size:
2 sets
5 sets
7 sets
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18779667 5 Plex 5 sets
18730749 7 Plex 7 sets
18769558 2 Plex 2 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18779667 Supplier R&D Systems™ Supplier No. FCSTM1305
  • ONLINE EXCLUSIVE PRICE
    2175.00€
    2052.75€ / 5 sets
    Save 122.25€
    5% Off
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Earn double rewards for reviews
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 Plex
Species Human
Binding Target Multiplex Cytokine Assay Kits
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.