Learn More
R&D Systems Human Biomarker Premixed Magnetic Luminex™ Performance Assay Kit A
Product Code. 30002081
Change view
Click to view available options
Quantity:
3 Plex
4 Plex
6 Plex
8 Plex
9 Plex
Unit Size:
3 sets
4 sets
6 sets
8 sets
9 sets
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30002081 4 Plex 4 sets
30002083 3 Plex 3 sets
30002076 9 Plex 9 sets
30002077 8 Plex 8 sets
30002080 6 Plex 6 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30002081 Supplier R&D Systems Supplier No. FCSTM1304
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 4 Plex
Species Human
Binding Target Multiplex Cytokine Assay Kits
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.