missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BCL6B Partial ORF (NP_862827, 248 a.a. - 345 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00255877-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192617

  • 528.00€ / 25µg

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

Sequence: GPIPGPQSRLSPTAATVQFKCGAPASTPYLLTSQAQDTSGSPSERARPLPGSEFFSCQNCEAVAGCSSGLDSLVPGDEDKPYKCQLCRSSFRYKGNLA
Specifications

Specifications

NP_862827
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GPIPGPQSRLSPTAATVQFKCGAPASTPYLLTSQAQDTSGSPSERARPLPGSEFFSCQNCEAVAGCSSGLDSLVPGDEDKPYKCQLCRSSFRYKGNLA
RUO
BCL6B
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
255877
BCL6B (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BAZF/ZBTB28/ZNF62
BCL6B
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers