Learn More
Abnova™ Human BCL11A Full-length ORF (NP_612569.1, 1 a.a. - 243 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00053335-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a C2H2 type zinc-finger protein by its similarity to the mouse Bcl11a/Evi9 protein. The corresponding mouse gene is a common site of retroviral integration in myeloid leukemia, and may function as a leukemia disease gene, in part, through its interaction with BCL6. During hematopoietic cell differentiation, this gene is down-regulated. It is possibly involved in lymphoma pathogenesis since translocations associated with B-cell malignancies also deregulates its expression. Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MSRRKQGKPQHLSKREFSPEPLEAILTDDEPDHGPLGAPEGDHDLLTCGQCQMNFPLGDILIFIEHKRKQCNGSLCLEKAVDKPPSPSPIEMKKASNPVEVGIQVTPEDDDCLSTSSRGICPKQEHIADKLLHWRGLSSPRSAHGALIPTPGMSAEYAPQGICKDEPSSYTCTTCKQPFTSAWFLLQHAQNTHGLRIYLESEHGSPLTPRVLHTPPFGVVPRELKMCGSFRMEAREPLSSEKISpecifications
NP_612569.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
53.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
BCL11A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
53335 | |
BCL11A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BCL11A-L/BCL11A-S/BCL11A-XL/CTIP1/EVI9/FLJ10173/FLJ34997/HBFQTL5/KIAA1809/ZNF856 | |
BCL11A | |
Recombinant | |
wheat germ expression system |