missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BCKDK Partial ORF (AAH07363, 31 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010295-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16164852

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Sequence: STSATDTHHVEMARERSKTVTSFYNQSAIDAAAEKPSVRLTPTMMLYAGRSQDGSHLLKSARYLQQELPVRIAHRIKGFRCLPFIIGCNP
Specifications

Specifications

AAH07363
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.53kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
STSATDTHHVEMARERSKTVTSFYNQSAIDAAAEKPSVRLTPTMMLYAGRSQDGSHLLKSARYLQQELPVRIAHRIKGFRCLPFIIGCNP
RUO
BCKDK
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
10295
BCKDK (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BCKDK
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers